Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(-)-1-Boc-3-(aminomethyl)pyrrolidine, 97%, Thermo Scientific™

Catalog No. AAH66277MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66277MD 250 mg
AAH6627703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH66277MD Supplier Thermo Scientific Chemicals Supplier No. H66277MD

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 199174-29-3
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.282
MDL Number MFCD03844734
InChI Key OGCCBDIYOAFOGK-MRVPVSSYSA-N
Synonym r-1-boc-3-aminomethyl pyrrolidine, r-n-boc-3-aminomethyl pyrrolidine, r-tert-butyl 3-aminomethyl pyrrolidine-1-carboxylate, r-3-aminomethyl-1-n-boc-pyrrolidine, tert-butyl 3r-3-aminomethyl pyrrolidine-1-carboxylate, r-1-n-boc-3-aminomethyl pyrrolidine, r-1-boc-3-aminomethylpyrrolidine, 3r-3-aminomethyl-1-boc-pyrrolidine, r---1-boc-3-aminomethyl pyrrolidine
PubChem CID 1515152
IUPAC Name tert-butyl (3R)-3-(aminomethyl)pyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(C1)CN

Specifications

Color Colorless to Yellow
Refractive Index 1.478
Quantity 250 mg
Beilstein 10537871
Sensitivity Air sensitive
Formula Weight 200.28
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material (R)-(-)-1-Boc-3-(aminomethyl)pyrrolidine
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold; Store under Nitrogen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.