Learn More
Medchemexpress LLC (R)-1-Boc-3-methyl-piperazine | 163765-44-4 | 99.9% | 200.28 | 25 G
SDP
Catalog No. 5000403098
Encompass_Preferred
The product (R)-1-Boc-3-methyl-piperazine is a chemical intermediate used for the preparation of Compound I3. Compound I3 exhibits anticancer activity against colorectal adenocarcinoma with BRCA2 gene deletion.
  • Used for the preparation of compound I3
  • Exhibits anticancer activity against colorectal adenocarcinoma with BRCA2 gene deletion
  • Appears as a solid below 40°C and a liquid above 45°C
  • Colorless to off-white in appearance
  • Recommended storage at 4°C, protected from light
Catalog No. 50-004-03098 Supplier Medchemexpress LLC Supplier No. 25GHY40041
The product (R)-1-Boc-3-methyl-piperazine is a chemical intermediate used for the preparation of Compound I3. Compound I3 exhibits anticancer activity against colorectal adenocarcinoma with BRCA2 gene deletion.
  • Used for the preparation of compound I3
  • Exhibits anticancer activity against colorectal adenocarcinoma with BRCA2 gene deletion
  • Appears as a solid below 40°C and a liquid above 45°C
  • Colorless to off-white in appearance
  • Recommended storage at 4°C, protected from light