Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-1-Ethyl-2-pyrrolidinecarboxamide, TCI America™
SDP

Catalog No. E04681G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E04681G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E04681G Supplier TCI America Supplier No. E04681G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 381670-32-2
Molecular Formula C7H14N2O
Molecular Weight (g/mol) 142.202
MDL Number MFCD00671532
InChI Key LQOASEHNECNLNM-ZCFIWIBFSA-N
Synonym N-Ethyl-D-prolinamide
PubChem CID 22215364
IUPAC Name (2R)-1-ethylpyrrolidine-2-carboxamide
SMILES CCN1CCCC1C(=O)N

Specifications

Melting Point 111°C
Color White
Quantity 1 g
Formula Weight 142.20
Physical Form Crystalline Powder
Chemical Name or Material (R)-(+)-1-Ethyl-2-pyrrolidinecarboxamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.