Learn More
Medchemexpress LLC R-1 Methanandamide Phosphate | 649569-33-5 | 98.0% | 1 ML
SDP
Catalog No. 5000342069
Encompass_Preferred
R-1 Methanandamide Phosphate is a water-soluble prodrug analog of arachidonylethanolamide (AEA), an endogenous cannabinoid compound. It is intended for research use only and not for patient use.
  • Water-soluble prodrug analog of arachidonylethanolamide (AEA)
  • Endogenous cannabinoid compound
  • Appears as a viscous liquid
  • Yellow to brown color
  • For research use only
  • Store at -20°C, protected from light
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light
Catalog No. 50-003-42069 Supplier Medchemexpress LLC Supplier No. HY12671910MM/1ML
R-1 Methanandamide Phosphate is a water-soluble prodrug analog of arachidonylethanolamide (AEA), an endogenous cannabinoid compound. It is intended for research use only and not for patient use.
  • Water-soluble prodrug analog of arachidonylethanolamide (AEA)
  • Endogenous cannabinoid compound
  • Appears as a viscous liquid
  • Yellow to brown color
  • For research use only
  • Store at -20°C, protected from light
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light