Learn More
Medchemexpress LLC R-1 methanandamide phosphate | 649569-33-5 | MFCD08062135 | >=98.0% | 441.54 g·mol⁻¹ | C23H40NO5P | 5 MG
SDP
Catalog No. 5000263886
Encompass_Preferred
R-1 Methanandamide Phosphate is a water-soluble phosphate prodrug analog of arachidonylethanolamide (AEA), an endogenous cannabinoid. Provided for research use, it enables aqueous handling and delivery compared with the parent lipid AEA and is supplied both as a solid and as ready-to-use DMSO solution for biochemical and cellular studies.
  • Water-soluble phosphate prodrug of arachidonylethanolamide (AEA).
  • Facilitates aqueous handling and administration compared with lipid AEA.
  • Suitable for in vitro research on cannabinoid receptor signaling.
  • Available as powder and as 10 mM solution in DMSO.
  • High purity reported (≥98%).
  • Molecular weight 441.54 g·mol⁻¹; formula C23H40NO5P.
Catalog No. 50-002-63886 Supplier Medchemexpress LLC Supplier No. HY1267195MG
R-1 Methanandamide Phosphate is a water-soluble phosphate prodrug analog of arachidonylethanolamide (AEA), an endogenous cannabinoid. Provided for research use, it enables aqueous handling and delivery compared with the parent lipid AEA and is supplied both as a solid and as ready-to-use DMSO solution for biochemical and cellular studies.
  • Water-soluble phosphate prodrug of arachidonylethanolamide (AEA).
  • Facilitates aqueous handling and administration compared with lipid AEA.
  • Suitable for in vitro research on cannabinoid receptor signaling.
  • Available as powder and as 10 mM solution in DMSO.
  • High purity reported (≥98%).
  • Molecular weight 441.54 g·mol⁻¹; formula C23H40NO5P.