Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (R)-1-N-Boc-4-N-Fmoc-2-piperazine carboxylic acid

Catalog No. CDS015197 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS015197 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS015197 Supplier Sigma Aldrich Supplier No. CDS015197100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Trimethyl(propargyl)silane (2-propynyl-trimethylsilane) is a clear colorless liquid and contains 500ppm of butylated hydroxytoluene (bht) as stabilizer
  • It undergoes gacl3-catalyzed dehydrogenation reaction with aldimines to afford 4-methylquinoline.trimethyl(propargyl)silane is suitable for use in the synthesis of diverse end-functionalized (-d-carboxyl, -d-hydroxy, -d-methyl-vinyl, -d-trimethylsilane, and -d-glycidyl-ether) via ααclickαα reaction
  • It may be used in the synthesis of α-allenyl-gaba (α-aminobutyric acid), via lewis acid mediated reaction with -d-ethoxy lactams

Specifications

Molecular Formula C25H28N2O6
Linear Formula C25H28N2O6
MDL Number MFCD02179102
Quantity 1 g
Molecular Weight (g/mol) 452.5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.