Learn More
Medchemexpress LLC (R)-1-(Naphthalen-1-yl)ethanamine hydrochloride | 82572-04-1 | 99.8% | C₁₂H₁₄ClN | 1 G
SDP
Catalog No. 5000434496
Encompass_Preferred
(R)-1-(Naphthalen-1-yl)ethanamine hydrochloride is a white to off-white solid primarily intended for research use only and has not been fully validated for medical applications.
  • Consistent with structure as verified by ¹H NMR spectrum and LCMS.
  • High purity of 99.84% (LCMS).
  • Store at 4°C under nitrogen.
  • For solvent solutions, store at -80°C for 6 months or -20°C for 1 month under nitrogen.
Catalog No. 50-004-34496 Supplier Medchemexpress LLC Supplier No. HY769651G
(R)-1-(Naphthalen-1-yl)ethanamine hydrochloride is a white to off-white solid primarily intended for research use only and has not been fully validated for medical applications.
  • Consistent with structure as verified by ¹H NMR spectrum and LCMS.
  • High purity of 99.84% (LCMS).
  • Store at 4°C under nitrogen.
  • For solvent solutions, store at -80°C for 6 months or -20°C for 1 month under nitrogen.