Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-1-(Naphthalen-1-yl)ethanamine hydrochloride | 82572-04-1 | 99.8% | C12H14CIN | 500 MG
SDP

Catalog No. 5000434467
Encompass_Preferred

The (R)-1-(Naphthalen-1-yl)ethanamine hydrochloride is a chemical compound supplied as a white to off-white solid. With a high purity of 99.8% as confirmed by LCMS, this substance is suitable for various research and laboratory applications.

  • White to off-white solid appearance
  • Purity of 99.8% (LCMS)
  • 1H NMR spectrum consistent with structure
  • LCMS consistent with structure
  • Store at 4°C under nitrogen
  • Store in solvent at -80°C for 6 months or -20°C for 1 month under nitrogen

Catalog No. 50-004-34467 Supplier Medchemexpress LLC Supplier No. HY76965500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The (R)-1-(Naphthalen-1-yl)ethanamine hydrochloride is a chemical compound supplied as a white to off-white solid. With a high purity of 99.8% as confirmed by LCMS, this substance is suitable for various research and laboratory applications.

  • White to off-white solid appearance
  • Purity of 99.8% (LCMS)
  • 1H NMR spectrum consistent with structure
  • LCMS consistent with structure
  • Store at 4°C under nitrogen
  • Store in solvent at -80°C for 6 months or -20°C for 1 month under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.