Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-1-(Naphthalen-1-yl)ethanamine hydrochloride | 82572-04-1 | 99.8% | C12H14ClN | 250 MG
SDP

Catalog No. 5000358583
Encompass_Preferred

(R)-1-(Naphthalen-1-yl)ethanamine hydrochloride is a high-purity chemical compound with the molecular formula C12H14ClN and a molecular weight of 207.70. It is typically supplied as a white to off-white solid.

  • Purity of 99.84% (LCMS) ensures reliable experimental results.
  • Product has been tested and complies with given specifications.
  • Recommended storage at 4°C under nitrogen to maintain stability.
  • For storage in solvent, maintain at -80°C for up to 6 months or -20°C for 1 month, both under nitrogen.

Catalog No. 50-003-58583 Supplier Medchemexpress LLC Supplier No. 250MGHY76965
May include imposed supplier surcharges.
Add to Cart
Add to Cart

(R)-1-(Naphthalen-1-yl)ethanamine hydrochloride is a high-purity chemical compound with the molecular formula C12H14ClN and a molecular weight of 207.70. It is typically supplied as a white to off-white solid.

  • Purity of 99.84% (LCMS) ensures reliable experimental results.
  • Product has been tested and complies with given specifications.
  • Recommended storage at 4°C under nitrogen to maintain stability.
  • For storage in solvent, maintain at -80°C for up to 6 months or -20°C for 1 month, both under nitrogen.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.