Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R-(+)-1-Phenylethanol, 99%, Thermo Scientific™

Catalog No. AAB2116006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2116006 5 g
AAB2116003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2116006 Supplier Thermo Scientific Chemicals Supplier No. B2116006

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1517-69-7
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00064263
InChI Key WAPNOHKVXSQRPX-UHFFFAOYNA-N
Synonym r-1-phenylethanol, r-+-1-phenylethanol, 1r-1-phenylethanol, r-alpha-methylbenzyl alcohol, r-+-sec-phenethyl alcohol, 1r-1-phenylethan-1-ol, r-+-1-phenylethyl alcohol, 1-phenylethanol, r, benzenemethanol, alpha-methyl-, r
PubChem CID 637516
ChEBI CHEBI:45616
IUPAC Name 1-phenylethan-1-ol
SMILES CC(O)C1=CC=CC=C1

Specifications

Boiling Point 97°C to 99°C (20 mmHg)
Quantity 5 g
Formula Weight 122.17
Percent Purity 99%
Chemical Name or Material R-(+)-1-Phenylethanol
Hazard Category Danger
Hazard Statement GHS H Statement
H301-H341-H227-H315-H319-H335
Toxic if swallowed.
Suspected of causing genetic defects.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P210-P261-P301+P310-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.