Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R-(+)-1-Phenylethanol, 99%, Thermo Scientific™

Catalog No. AAB2116003
Click to view available options
Quantity:
1 g
5 g

Catalog No. AAB2116003

Supplier: Thermo Scientific Chemicals B2116003

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1517-69-7
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00064263
InChI Key WAPNOHKVXSQRPX-UHFFFAOYNA-N
Synonym r-1-phenylethanol, r-+-1-phenylethanol, 1r-1-phenylethanol, r-alpha-methylbenzyl alcohol, r-+-sec-phenethyl alcohol, 1r-1-phenylethan-1-ol, r-+-1-phenylethyl alcohol, 1-phenylethanol, r, benzenemethanol, alpha-methyl-, r
PubChem CID 637516
ChEBI CHEBI:45616
IUPAC Name 1-phenylethan-1-ol
SMILES CC(O)C1=CC=CC=C1

Specifications

Boiling Point 97°C to 99°C (20 mmHg)
Quantity 1 g
Formula Weight 122.17
Percent Purity 99%
Chemical Name or Material R-(+)-1-Phenylethanol
Hazard Category Danger
Hazard Statement GHS H Statement
H301-H341-H227-H315-H319-H335
Toxic if swallowed.
Suspected of causing genetic defects.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P210-P261-P301+P310-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.