Learn More
Strem, An Ascensus Company CAS# 565184-33-0. 500mg. (R)-(-)-1-[(R)-2-(2'-Diphenylphosphinophenyl)ferrocenyl]ethylbis(di-3,5-trifluoromethylphenyl)phosphine, min. 97%. MFCD04112740
SDP
Supplier:  Strem, An Ascensus Company 261300500MG
Encompass
CAS# 565184-33-0. 500mg. (R)-(-)-1-[(R)-2-(2'-Diphenylphosphinophenyl)ferrocenyl]ethylbis(di-3,5-trifluoromethylphenyl)phosphine, min. 97%. MFCD04112740. Molecular Weight: 930.52. Molecular Formula: C46H32F12FeP2. Color/form: orange pwdr. Strem# 26-1300.
Catalog No. 50-901-11048