Learn More
Strem, An Ascensus Company CAS# 155830-69-6. 100mg. (R)-(-)-1-[(S)-2-(Diphenylphosphino)ferrocenyl]ethyldi-t-butylphosphine, min. 97%. MFCD01074442
SDP
Supplier:  Strem, An Ascensus Company 261200100MG
Encompass
CAS# 155830-69-6. 100mg. (R)-(-)-1-[(S)-2-(Diphenylphosphino)ferrocenyl]ethyldi-t-butylphosphine, min. 97%. MFCD01074442. Molecular Weight: 542.46. Molecular Formula: C32H40FeP2. Color/form: orange pwdr. Strem# 26-1200. http://www.strem.com/catalog/v/26-1200/
Catalog No. 50-901-11035