Learn More
Strem, An Ascensus Company CAS# 184095-69-0. 500mg. (R)-(-)-1-[(S)-2-(Diphenylphosphino)ferrocenyl]ethyldi-3,5-xylylphosphine, min. 97%. MFCD03426997
SDP
Supplier:  Strem, An Ascensus Company 261255500MG
Encompass
CAS# 184095-69-0. 500mg. (R)-(-)-1-[(S)-2-(Diphenylphosphino)ferrocenyl]ethyldi-3,5-xylylphosphine, min. 97%. MFCD03426997. Molecular Weight: 638.56. Molecular Formula: C40H40FeP2. Color/form: orange pwdr. Strem# 26-1255. http://www.strem.com/catalog/v/26-1255/
Catalog No. 50-901-11046