Learn More
Strem, An Ascensus Company CAS# 155806-35-2. 500mg. (R)-(-)-1-[(S)-2-(Diphenylphosphino)ferrocenyl]ethyldicyclohexylphosphine ethanol adduct, min. 97% (R)-(S)-JOSIPHOS. MFCD00800284
SDP
Catalog No. 5090111040
Encompass
CAS# 155806-35-2. 500mg. (R)-(-)-1-[(S)-2-(Diphenylphosphino)ferrocenyl]ethyldicyclohexylphosphine ethanol adduct, min. 97% (R)-(S)-JOSIPHOS. MFCD00800284. Molecular Weight: 594.59 (640.66). Molecular Formula: C36H44FeP2∙CH3CH2OH. Color/form: orange pwdr. Strem# 26-1210. http://www.strem.com/catalog/v/26-1210/
Catalog No. 50-901-11040 Supplier Strem, An Ascensus Company Supplier No. 261210500MG
CAS# 155806-35-2. 500mg. (R)-(-)-1-[(S)-2-(Diphenylphosphino)ferrocenyl]ethyldicyclohexylphosphine ethanol adduct, min. 97% (R)-(S)-JOSIPHOS. MFCD00800284. Molecular Weight: 594.59 (640.66). Molecular Formula: C36H44FeP2∙CH3CH2OH. Color/form: orange pwdr. Strem# 26-1210. http://www.strem.com/catalog/v/26-1210/