Learn More
Strem, An Ascensus Company CAS# 158923-11-6. (R)-(-)-1-[(S)-2-(Dicyclohexylphosphino)ferrocenyl]ethyldi-t-butylphosphine, min. 97%. MFCD08543435.100MG
SDP
Supplier:  Strem, An Ascensus Company 260975100MG
Encompass
CAS# 158923-11-6. 100MG. (R)-(-)-1-[(S)-2-(Dicyclohexylphosphino)ferrocenyl]ethyldi-t-butylphosphine, min. 97%. MFCD08543435. Molecular Formula: C32H52FeP2. Molecular Weight: 554.56. Color/Form: orange pwdr. Strem#: 26-0975 www.strem.com/catalog/v3/26-0975/
Catalog No. 50-138-5459