Learn More
Selleck Chemical LLC R-10015
SDP
Catalog No. 5000002847
Encompass_Preferred
R-10015 is a potent selective inhibitor of LIM domain kinase (LIMK) with IC50 of 38 nM for human LIMK1 R-10015 binds to the ATP-binding pocket and acts as a broad-spectrum antiviral compound for HIV infection
Catalog No. 50-000-02847 Supplier Selleck Chemical LLC Supplier No. S68372MG
R-10015 is a potent selective inhibitor of LIM domain kinase (LIMK) with IC50 of 38 nM for human LIMK1 R-10015 binds to the ATP-binding pocket and acts as a broad-spectrum antiviral compound for HIV infection