Learn More
Medchemexpress LLC HY-120097 5mg Medchemexpress, R-10015 CAS:2097938-51-5 Purity:>98%
SDP
Catalog No. 501932548
Encompass_Preferred
Medchemexpress, HY-120097 5mg R-10015 CAS:2097938-51-5 R-10015, a broad-spectrum antiviral compound for HIV infection, acts as a potent and selective inhibitor of LIM domain kinase (LIMK) and binds to the ATP-binding pocket, with an IC 50 of 38 nM for human LIMK1 [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2548 Supplier Medchemexpress LLC Supplier No. HY1200975MG
Medchemexpress, HY-120097 5mg R-10015 CAS:2097938-51-5 R-10015, a broad-spectrum antiviral compound for HIV infection, acts as a potent and selective inhibitor of LIM domain kinase (LIMK) and binds to the ATP-binding pocket, with an IC 50 of 38 nM for human LIMK1 [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.