Learn More
Medchemexpress LLC HY-Z0031 500mg Medchemexpress, (R)-(-)-1,2-Propanediol CAS:4254-14-2 Purity:>98%
Supplier: Medchemexpress LLC HYZ0031500MG
Medchemexpress, HY-Z0031 500mg (R)-(-)-1,2-Propanediol CAS:4254-14-2 (R)-(-)-1,2-Propanediol is a (R)-enantiomer of 1,2-Propanediol that produced from glucose in Escherichia coli expressing NADH-linked glycerol dehydrogenase genes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.