Learn More
Medchemexpress LLC HY-10444 5mg Medchemexpress, R-1479 CAS:478182-28-4 Purity:>98%
SDP
Catalog No. 501968222
Encompass_Preferred
Medchemexpress, HY-10444 5mg R-1479 CAS:478182-28-4 R-1479 is a specific inhibitor of HCV replication in the HCV subgenomic replicon system (IC50=1.28 μM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8222 Supplier Medchemexpress LLC Supplier No. HY104445MG
Medchemexpress, HY-10444 5mg R-1479 CAS:478182-28-4 R-1479 is a specific inhibitor of HCV replication in the HCV subgenomic replicon system (IC50=1.28 μM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.