Learn More
Medchemexpress LLC (R,1R)-Tenofovir amibufenamide | 1571076-37-3 | 98.9% | 490.49 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY137453C1MG
Encompass_Preferred
(R,1R)-Tenofovir amibufenamide can be used for purifying a tenofovir prodrug. Tenofovir is a nucleoside acids reverse transcriptase inhibitor. This compound appears as a white to off-white solid.
  • Used for purifying a tenofovir prodrug
  • Functions as a nucleoside acids reverse transcriptase inhibitor
  • White to off-white solid appearance
Catalog No. 50-003-52264