Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R,1R)-Tenofovir amibufenamide | 1571076-37-3 | 98.9% | 490.49 | 25 MG
SDP

Catalog No. 5000351012
Encompass_Preferred

(R,1R)-Tenofovir amibufenamide can be used for purifying a tenofovir prodrug. Tenofovir is a nucleoside acids reverse transcriptase inhibitor.

  • Used for purifying a tenofovir prodrug
  • Functions as a nucleoside acids reverse transcriptase inhibitor
  • High purity of 98.9%
  • Molecular weight of 490.49
  • Appears as a white to off-white solid
  • Recommended storage at 4°C under nitrogen

Catalog No. 50-003-51012 Supplier Medchemexpress LLC Supplier No. 25MGHY137453C
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R,1R)-Tenofovir amibufenamide can be used for purifying a tenofovir prodrug. Tenofovir is a nucleoside acids reverse transcriptase inhibitor.

  • Used for purifying a tenofovir prodrug
  • Functions as a nucleoside acids reverse transcriptase inhibitor
  • High purity of 98.9%
  • Molecular weight of 490.49
  • Appears as a white to off-white solid
  • Recommended storage at 4°C under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.