Learn More
Medchemexpress LLC (R)-2-(1-(((9H-Fluoren-9-yl)methoxy)carbonyl)pyrrolidin-2-yl)acetic acid | 193693-61-7 | 100.0% | 351.40 g/mol | C21H21NO4 | 1 G
SDP
Catalog No. 5000210226
Encompass_Preferred
(R)-2-(1-(((9H-Fluoren-9-yl)methoxy)carbonyl)pyrrolidin-2-yl)acetic acid is an Fmoc-protected proline derivative supplied as a high-purity research reagent intended for biochemical synthesis and analytical applications. It is provided as the R-enantiomer and is for research use only.
  • Fmoc-protected proline derivative.
  • Chiral R-enantiomer.
  • High purity (99.96%).
  • Molecular weight 351.40 g/mol and formula C21H21NO4.
  • Available in 1 G, 5 G, 10 G, and 50 G pack sizes.
  • Intended for research use only; not for human or animal therapeutic use.
Catalog No. 50-002-10226 Supplier Medchemexpress LLC Supplier No. HYW0452211G
(R)-2-(1-(((9H-Fluoren-9-yl)methoxy)carbonyl)pyrrolidin-2-yl)acetic acid is an Fmoc-protected proline derivative supplied as a high-purity research reagent intended for biochemical synthesis and analytical applications. It is provided as the R-enantiomer and is for research use only.
  • Fmoc-protected proline derivative.
  • Chiral R-enantiomer.
  • High purity (99.96%).
  • Molecular weight 351.40 g/mol and formula C21H21NO4.
  • Available in 1 G, 5 G, 10 G, and 50 G pack sizes.
  • Intended for research use only; not for human or animal therapeutic use.