Learn More
Medchemexpress LLC (R)-2-(1-(((9H-Fluoren-9-yl)methoxy)carbonyl)pyrrolidin-2-yl)acetic acid | 193693-61-7 | 100.0% | 351.40 g/mol | C21H21NO4 | 5 G
SDP
Catalog No. 5000210063
Encompass_Preferred
(R)-2-(1-(((9H-Fluoren-9-yl)methoxy)carbonyl)pyrrolidin-2-yl)acetic acid is an Fmoc-protected proline/pyrrolidine-derived acetic acid supplied as a high-purity research reagent. It is used as a building block in peptide synthesis and as an intermediate in organic synthesis for medicinal chemistry research.
  • High purity suitable for synthesis and analytical work.
  • Fmoc-protected proline derivative for Fmoc-based solid-phase peptide synthesis.
  • Suitable as a synthetic intermediate in medicinal chemistry and SAR studies.
  • Available in multiple bench-scale pack sizes for laboratory use.
  • Comes with SDS and certificate of analysis for handling and quality verification.
Catalog No. 50-002-10063 Supplier Medchemexpress LLC Supplier No. HYW0452215G
(R)-2-(1-(((9H-Fluoren-9-yl)methoxy)carbonyl)pyrrolidin-2-yl)acetic acid is an Fmoc-protected proline/pyrrolidine-derived acetic acid supplied as a high-purity research reagent. It is used as a building block in peptide synthesis and as an intermediate in organic synthesis for medicinal chemistry research.
  • High purity suitable for synthesis and analytical work.
  • Fmoc-protected proline derivative for Fmoc-based solid-phase peptide synthesis.
  • Suitable as a synthetic intermediate in medicinal chemistry and SAR studies.
  • Available in multiple bench-scale pack sizes for laboratory use.
  • Comes with SDS and certificate of analysis for handling and quality verification.