Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-2-(1-(((9H-fluoren-9-yl)methoxy)carbonyl)pyrrolidin-2-yl)acetic acid | 193693-61-7 | MFCD06202403 | 100.0% | 351.40 g/mol | C21H21NO4 | 10 G
SDP

Catalog No. 5000210202
Encompass_Preferred

(R)-2-(1-(((9H-fluoren-9-yl)methoxy)carbonyl)pyrrolidin-2-yl)acetic acid is an Fmoc-protected proline derivative used as a building block in peptide synthesis and other research applications. It is supplied for research use only and is not intended for human or clinical use.

  • Fmoc-protected proline derivative suitable for peptide synthesis
  • Molecular formula C21H21NO4
  • Molecular weight 351.40 g/mol
  • High purity (manufacturer reports 99.96%)
  • Available in multiple sizes, including 1 g, 5 g, 10 g, and 50 g
  • Provided for research use only; follow laboratory safety and handling procedures

Catalog No. 50-002-10202 Supplier Medchemexpress LLC Supplier No. HYW04522110G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-2-(1-(((9H-fluoren-9-yl)methoxy)carbonyl)pyrrolidin-2-yl)acetic acid is an Fmoc-protected proline derivative used as a building block in peptide synthesis and other research applications. It is supplied for research use only and is not intended for human or clinical use.

  • Fmoc-protected proline derivative suitable for peptide synthesis
  • Molecular formula C21H21NO4
  • Molecular weight 351.40 g/mol
  • High purity (manufacturer reports 99.96%)
  • Available in multiple sizes, including 1 g, 5 g, 10 g, and 50 g
  • Provided for research use only; follow laboratory safety and handling procedures

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.