Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-2-(1-Hydroxyethyl)pyridine

Catalog No. AAJ6254803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6254803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6254803 Supplier Thermo Scientific Chemicals Supplier No. J6254803
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 27911-63-3 | C7H9NO | 123.155 g/mol

For synthesis of optically active products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
For synthesis of optically active products.

Solubility
Soluble in ethanol and water.

Notes
Store in a tightly closed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27911-63-3
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
MDL Number MFCD04972322
InChI Key PPHIIIRFJKDTLG-ZCFIWIBFSA-N
Synonym r-1-pyridin-2-yl ethanol, r-2-1-hydroxyethyl pyridine, r-1-2-pyridyl ethanol, 1r-1-pyridin-2-yl ethan-1-ol, r-alpha-methyl-2-pyridinemethanol, 1r-1-pyridin-2-ylethanol, 1r-1-pyridin-2-yl ethanol, pubchem5699, r-1-pyridin-2-yl-ethanol, 1r-1-2-pyridyl ethanol
PubChem CID 642847
IUPAC Name (1R)-1-pyridin-2-ylethanol
SMILES CC(C1=CC=CC=N1)O

Specifications

Color Colorless
Quantity 1 g
Beilstein 3537186
Solubility Information Soluble in ethanol and water.
Formula Weight 123.16
Physical Form Solid
Chemical Name or Material (R)-2-(1-Hydroxyethyl)pyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.