Learn More
Medchemexpress LLC (R)-2-(2,2-diphenylacetamido)-5-guanidino-N-((R)-1-(4-hydroxyphenyl)ethyl)pentanamide formate | 00-00-0 | MFCD04112988 | 99.8% | 533.62 g/mol | C29H35N5O5 | 25 MG
SDP
Catalog No. 5000249055
Encompass_Preferred
Y1 receptor antagonist 1 (formic) is a research-grade small molecule antagonist of neuropeptide Y Y1 receptors supplied as the formate salt. It is intended for in vitro and preclinical studies of GPCR-mediated neuronal and cardiovascular pathways. Manufacturer documentation reports molecular formula C29H35N5O5, molecular weight 533.62 g/mol, and high purity (LCMS 99.84%).
  • High purity (LCMS) 99.8%.
  • Formate salt with molecular formula C29H35N5O5.
  • Molecular weight 533.62 g/mol.
  • Supplied in 25 MG quantities suitable for small-scale research.
  • Store sealed, away from moisture; solid at -20°C; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Suitable for in vitro and preclinical GPCR and neuronal signaling studies.
Catalog No. 50-002-49055 Supplier Medchemexpress LLC Supplier No. HY101704A25MG
Y1 receptor antagonist 1 (formic) is a research-grade small molecule antagonist of neuropeptide Y Y1 receptors supplied as the formate salt. It is intended for in vitro and preclinical studies of GPCR-mediated neuronal and cardiovascular pathways. Manufacturer documentation reports molecular formula C29H35N5O5, molecular weight 533.62 g/mol, and high purity (LCMS 99.84%).
  • High purity (LCMS) 99.8%.
  • Formate salt with molecular formula C29H35N5O5.
  • Molecular weight 533.62 g/mol.
  • Supplied in 25 MG quantities suitable for small-scale research.
  • Store sealed, away from moisture; solid at -20°C; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Suitable for in vitro and preclinical GPCR and neuronal signaling studies.