Learn More
Strem, An Ascensus Company CAS# 1365531-89-0. 100mg. (R)-2,2'-Bis[bis(3,5-dimethylphenyl)phosphino]-4,4',6,6'-tetramethoxy-)-1,1'-biphenyl, min. 97% (R)-Xyl-Garphos™. MFCD19443624
SDP
Supplier:  Strem, An Ascensus Company 151661100MG
Encompass
CAS# 1365531-89-0. 100mg. (R)-2,2'-Bis[bis(3,5-dimethylphenyl)phosphino]-4,4',6,6'-tetramethoxy-)-1,1'-biphenyl, min. 97% (R)-Xyl-Garphos™. MFCD19443624. Molecular Weight: 754.87. Molecular Formula: C48H52O4P2. Color/form: white xtl. Strem# 15-1661. http://www.strem.com/catalog/v/15-1661/
Catalog No. 50-370-0