Learn More
Strem, An Ascensus Company CAS# 394248-45-4. 500mg. (R)-(+)-2,2'-Bis[di(3,5-xylyl)phosphino]-6,6'-dimethoxy-1,1'-biphenyl, min. 97%. MFCD09753009
SDP
Catalog No. 5090109928
Encompass
CAS# 394248-45-4. 500mg. (R)-(+)-2,2'-Bis[di(3,5-xylyl)phosphino]-6,6'-dimethoxy-1,1'-biphenyl, min. 97%. MFCD09753009. Molecular Weight: 694.84. Molecular Formula: C46H48O2P2. Color/form: white pwdr. Strem# 15-0488. http://www.strem.com/catalog/v/15-0488
Catalog No. 50-901-09928 Supplier Strem, An Ascensus Company Supplier No. 150488500MG
CAS# 394248-45-4. 500mg. (R)-(+)-2,2'-Bis[di(3,5-xylyl)phosphino]-6,6'-dimethoxy-1,1'-biphenyl, min. 97%. MFCD09753009. Molecular Weight: 694.84. Molecular Formula: C46H48O2P2. Color/form: white pwdr. Strem# 15-0488. http://www.strem.com/catalog/v/15-0488