Learn More
Strem, An Ascensus Company CAS# 76189-55-4. 5g. (R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, 98% (R)-(+)-BINAP. MFCD00010805
SDP
Catalog No. 5090109794
Encompass
CAS# 76189-55-4. 5g. (R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, 98% (R)-(+)-BINAP. MFCD00010805. Molecular Weight: 622.70. Molecular Formula: C44H32P2. Color/form: white to off-white pwdr. Strem# 15-0150. http://www.strem.com/catalog/v/15-0150/
Catalog No. 50-901-09794 Supplier Strem, An Ascensus Company Supplier No. 1501505G
CAS# 76189-55-4. 5g. (R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, 98% (R)-(+)-BINAP. MFCD00010805. Molecular Weight: 622.70. Molecular Formula: C44H32P2. Color/form: white to off-white pwdr. Strem# 15-0150. http://www.strem.com/catalog/v/15-0150/