Learn More
Strem, An Ascensus Company CAS# 1365531-75-4. 100mg. (R)-2,2'-Bis(diphenylphosphino)-4,4',6,6'-tetramethoxy)-1,1'-biphenyl, min. 97% (R)-Ph-Garphos™. MFCD19443622
SDP
Supplier:  Strem, An Ascensus Company 151653100MG
Encompass
CAS# 1365531-75-4. 100mg. (R)-2,2'-Bis(diphenylphosphino)-4,4',6,6'-tetramethoxy)-1,1'-biphenyl, min. 97% (R)-Ph-Garphos™. MFCD19443622. Molecular Weight: 642.66. Molecular Formula: C40H36O4P2. Color/form: white xtl. Strem# 15-1653. http://www.strem.com/catalog/v/15-1653/
Catalog No. 50-369-2