Learn More
Strem, An Ascensus Company CAS# 173831-50-0. 5g. (R)-(+)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl, 98%. MFCD03788935
SDP
Catalog No. 5090109162
Encompass
CAS# 173831-50-0. 5g. (R)-(+)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl, 98%. MFCD03788935. Molecular Weight: 374.43. Molecular Formula: C24H22O4. Color/form: white pwdr. Strem# 08-0170. http://www.strem.com/catalog/v/08-0170/
Catalog No. 50-901-09162 Supplier Strem, An Ascensus Company Supplier No. 0801705G
CAS# 173831-50-0. 5g. (R)-(+)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl, 98%. MFCD03788935. Molecular Weight: 374.43. Molecular Formula: C24H22O4. Color/form: white pwdr. Strem# 08-0170. http://www.strem.com/catalog/v/08-0170/