Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-2,3-Di-(Boc-amino)propionic acid dicyclohexylammonium salt, 95%, Thermo Scientific™

Catalog No. AAH6228303
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6228303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6228303 Supplier Thermo Scientific Chemicals Supplier No. H6228303

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 201472-66-4
Molecular Formula C26H49N3O6
Molecular Weight (g/mol) 499.693
MDL Number MFCD00238312
InChI Key BGTQRELDRNOEDA-FVGYRXGTSA-N
Synonym boc-dab boc-oh.dcha, boc-dab boc-oh dcha, boc-dab boc-oh?cha, 2s-2,4-bis tert-butoxycarbonyl amino butanoic acid; dicha, 2s-2,4-bis tert-butoxycarbonyl amino butanoic acid n,n-dicyclohexylamine salt, 2s-2,4-bis tert-butoxycarbonyl amino butanoic acid n-cyclohexylcyclohexanamine, n-alpha-n-gamma-bis t-butyloxycarbonyl-l-2,4-diaminobutyric acid dicyclohexylamine
PubChem CID 56777023
IUPAC Name (2S)-2,4-bis[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid;N-cyclohexylcyclohexanamine
SMILES CC(C)(C)OC(=O)NCCC(C(=O)O)NC(=O)OC(C)(C)C.C1CCC(CC1)NC2CCCCC2

Specifications

Quantity 1 g
Formula Weight 485.66
Percent Purity 95%
Physical Form Solid
Chemical Name or Material (R)-2,3-Di-(Boc-amino)propionic acid dicyclohexylammonium salt
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.