Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(-)-2,3-Diaminopropionic Acid Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. D15731G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D15731G 1 g
D1573100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D15731G Supplier TCI America Supplier No. D15731G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6018-56-0
Molecular Formula C3H9ClN2O2
Molecular Weight (g/mol) 140.567
MDL Number MFCD00070606
InChI Key SKWCZPYWFRTSDD-HSHFZTNMSA-N
Synonym 3-amino-d-alanine hydrochloride, r-2,3-diaminopropanoic acid hydrochloride, h-d-dap-oh hcl, 2r-2,3-diaminopropanoic acid hydrochloride, unii-5452mgq13c, 3-amino-d-alanine monohydrochloride, r---2,3-diaminopropionic acid hydrochloride, d-alanine, 3-amino-, monohydrochloride, d-2,3-diaminopropionic acid hydrochloride, d-2,3-diaminopropionic acid monohydrochloride
PubChem CID 16218517
IUPAC Name (2R)-2,3-diaminopropanoic acid;hydrochloride
SMILES C(C(C(=O)O)N)N.Cl

Specifications

Melting Point 240°C
Color White-Yellow
Quantity 1 g
Formula Weight 140.57
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(-)-2,3-Diaminopropionic Acid Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.