Learn More
Medchemexpress LLC (R)-2,5-Dioxotetrahydrofuran-3-yl acetate | 79814-40-7 | 95.0% | C6H6O5 | 250 MG

Supplier: Medchemexpress LLC 250MGHY76985
(R)-2,5-Dioxotetrahydrofuran-3-yl acetate is a chemical compound that serves as a drug intermediate for the synthesis of various active compounds. This product is intended for research use only.
- Appearance: solid
- Molecular weight: 158.11
- Stable under recommended storage conditions
- Melting point: 56-58 °C
- Boiling point: 162 °C at 14 mmHg
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.