Learn More
Medchemexpress LLC (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2-(trifluoromethyl)phenyl)propanoic acid | 352523-15-0 | MFCD01863051 | 99.9% | 455.43 g·mol^-1 | C25H20F3NO4 | 500 MG
SDP
Catalog No. 5000282973
Encompass_Preferred
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2-(trifluoromethyl)phenyl)propanoic acid is an Fmoc-protected, optically active phenylalanine derivative intended for research and synthetic chemistry applications. It is characterized for peptide synthesis and related organic transformations, and is supplied with analytical documentation for laboratory use.
  • Fmoc-protected amino acid for solid-phase and solution-phase peptide synthesis.
  • Optically pure R-enantiomer to support stereospecific synthesis.
  • High purity, 99.9% (reported by RP-HPLC), suitable for research applications.
  • Molecular formula C25H20F3NO4 and molecular weight 455.43 g·mol^-1.
  • Available in pre-weighed laboratory pack sizes for convenient handling.
Catalog No. 50-002-82973 Supplier Medchemexpress LLC Supplier No. HYW130212500MG
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2-(trifluoromethyl)phenyl)propanoic acid is an Fmoc-protected, optically active phenylalanine derivative intended for research and synthetic chemistry applications. It is characterized for peptide synthesis and related organic transformations, and is supplied with analytical documentation for laboratory use.
  • Fmoc-protected amino acid for solid-phase and solution-phase peptide synthesis.
  • Optically pure R-enantiomer to support stereospecific synthesis.
  • High purity, 99.9% (reported by RP-HPLC), suitable for research applications.
  • Molecular formula C25H20F3NO4 and molecular weight 455.43 g·mol^-1.
  • Available in pre-weighed laboratory pack sizes for convenient handling.