Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(thiophen-3-yl)propanoic acid | 220497-90-5 | 99.4% | 393.46 | 1 G
SDP

Catalog No. 5000366286
Encompass_Preferred

(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(thiophen-3-yl)propanoic acid is a chemical compound primarily used as a laboratory chemical and for the manufacture of substances. It is intended for research use only and should be handled by suitably qualified and experienced scientists in appropriately equipped facilities.

  • Solid appearance
  • Molecular weight of 393.46
  • Purity of 99.4%
  • Recommended storage as powder at -20°C for 3 years or 4°C for 2 years
  • Recommended storage in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-003-66286 Supplier Medchemexpress LLC Supplier No. HYW0110031G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(thiophen-3-yl)propanoic acid is a chemical compound primarily used as a laboratory chemical and for the manufacture of substances. It is intended for research use only and should be handled by suitably qualified and experienced scientists in appropriately equipped facilities.

  • Solid appearance
  • Molecular weight of 393.46
  • Purity of 99.4%
  • Recommended storage as powder at -20°C for 3 years or 4°C for 2 years
  • Recommended storage in solvent at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.