Learn More
Medchemexpress LLC (R)-2-acetoxy-2-phenylacetic acid | 51019-43-3 | ≥97.0% | C10H10O4 | 1 KG
SDP
Catalog No. 5000432326
Encompass_Preferred
(R)-2-acetoxy-2-phenylacetic acid is a biochemical reagent suitable for use as a biological material or organic compound in life science-related research. Its (R)- and (S)-isomers also function as chiral derivatizing agents for determining the enantiomeric purity of α-deuterated carboxylic acids, alcohols, and amines via NMR.
  • Can be used as a biological material or organic compound for life science research.
  • The (R)- and (S)-isomers are effective for NMR determination of enantiomeric purity in α-deuterated carboxylic acids, alcohols, and amines.
Catalog No. 50-004-32326 Supplier Medchemexpress LLC Supplier No. HYA00251KG
(R)-2-acetoxy-2-phenylacetic acid is a biochemical reagent suitable for use as a biological material or organic compound in life science-related research. Its (R)- and (S)-isomers also function as chiral derivatizing agents for determining the enantiomeric purity of α-deuterated carboxylic acids, alcohols, and amines via NMR.
  • Can be used as a biological material or organic compound for life science research.
  • The (R)- and (S)-isomers are effective for NMR determination of enantiomeric purity in α-deuterated carboxylic acids, alcohols, and amines.