Learn More
Medchemexpress LLC (R)-2-acetoxy-2-phenylacetic acid | 51019-43-3 | ≥97.0% | C10H10O4 | 100 G
SDP
Catalog No. 5000433140
Encompass_Preferred
(R)-2-acetoxy-2-phenylacetic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
  • Used as a biochemical reagent
  • Utilized as a biological material or organic compound for life science related research
  • The (R)- and (S)-isomers serve as chiral derivatizing agents
  • Used for NMR determination of enantiomeric purity of α-deuterated carboxylic acids, alcohols, and amines
Catalog No. 50-004-33140 Supplier Medchemexpress LLC Supplier No. HYA0025100G
(R)-2-acetoxy-2-phenylacetic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
  • Used as a biochemical reagent
  • Utilized as a biological material or organic compound for life science related research
  • The (R)- and (S)-isomers serve as chiral derivatizing agents
  • Used for NMR determination of enantiomeric purity of α-deuterated carboxylic acids, alcohols, and amines