Learn More
Medchemexpress LLC (R)-2-Acetoxy-2-phenylacetic acid | 51019-43-3 | ≥97.0% | C10H10O4 | 50 G
SDP
Catalog No. 5000432487
Encompass_Preferred
(R)-2-acetoxy-2-phenylacetic acid is a biochemical reagent suitable for use as a biological material or organic compound in life science related research. It is intended for research use only.
  • Biochemical reagent for life science research
  • Used as a biological material or organic compound
  • Chiral derivatizing agent for NMR determination of enantiomeric purity
  • Solid, white to off-white appearance
  • Molecular weight: 194.19
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-004-32487 Supplier Medchemexpress LLC Supplier No. HYA002550G
(R)-2-acetoxy-2-phenylacetic acid is a biochemical reagent suitable for use as a biological material or organic compound in life science related research. It is intended for research use only.
  • Biochemical reagent for life science research
  • Used as a biological material or organic compound
  • Chiral derivatizing agent for NMR determination of enantiomeric purity
  • Solid, white to off-white appearance
  • Molecular weight: 194.19
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month