Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-2-amino-2-(4-fluorophenyl)acetic acid | 93939-74-3 | MFCD00042727 | 95.0% | 169.16 g/mol | C8H8FNO2 | 500 MG
SDP

Catalog No. 5000210924
Encompass_Preferred

(R)-2-amino-2-(4-fluorophenyl)acetic acid is the R-enantiomer of 4-fluorophenylglycine and is used as a chiral amino acid derivative for research and synthetic applications. It is supplied as a solid, white to off-white powder, with CAS 93939-74-3, purity of 95.0%, molecular formula C8H8FNO2, and molecular weight 169.16 g/mol. Store the powder as recommended for long-term stability.

  • Chiral R-enantiomer for stereospecific synthesis and studies.
  • Used as a building block in medicinal chemistry and peptide synthesis.
  • White to off-white solid suitable for analytical and synthetic work.
  • Purity 95.0% for reliable experimental use.
  • CAS 93939-74-3 for unambiguous chemical identification.

Catalog No. 50-002-10924 Supplier Medchemexpress LLC Supplier No. HYW051418500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-2-amino-2-(4-fluorophenyl)acetic acid is the R-enantiomer of 4-fluorophenylglycine and is used as a chiral amino acid derivative for research and synthetic applications. It is supplied as a solid, white to off-white powder, with CAS 93939-74-3, purity of 95.0%, molecular formula C8H8FNO2, and molecular weight 169.16 g/mol. Store the powder as recommended for long-term stability.

  • Chiral R-enantiomer for stereospecific synthesis and studies.
  • Used as a building block in medicinal chemistry and peptide synthesis.
  • White to off-white solid suitable for analytical and synthetic work.
  • Purity 95.0% for reliable experimental use.
  • CAS 93939-74-3 for unambiguous chemical identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.