Learn More
Medchemexpress LLC (R)-2-amino-2-(thiophen-3-yl)acetic acid | 1194-86-1 | MFCD00079614 | 60;98.0% | 157.19 g/mol | C6H7NO2S | 1 G
SDP
Catalog No. 5000299551
Encompass_Preferred
(R)-2-Amino-2-(thiophen-3-yl)acetic acid is the R-enantiomer of a thiophene-substituted glycine derivative provided as a white to off-white solid for research use. It functions as a chiral amino acid building block for medicinal chemistry, peptide synthesis, and structure-activity relationship studies, with reported purity ≥98.0% and molecular weight 157.19 g/mol.
  • Chiral R-enantiomer suitable for stereospecific synthesis.
  • High purity (≥98.0%) for reliable experimental results.
  • Solid form simplifies handling and storage.
  • Small molecular weight for incorporation into peptides and small molecules.
  • Thiophene substitution provides chemical diversity for SAR exploration.
Catalog No. 50-002-99551 Supplier Medchemexpress LLC Supplier No. HYW0172541G
(R)-2-Amino-2-(thiophen-3-yl)acetic acid is the R-enantiomer of a thiophene-substituted glycine derivative provided as a white to off-white solid for research use. It functions as a chiral amino acid building block for medicinal chemistry, peptide synthesis, and structure-activity relationship studies, with reported purity ≥98.0% and molecular weight 157.19 g/mol.
  • Chiral R-enantiomer suitable for stereospecific synthesis.
  • High purity (≥98.0%) for reliable experimental results.
  • Solid form simplifies handling and storage.
  • Small molecular weight for incorporation into peptides and small molecules.
  • Thiophene substitution provides chemical diversity for SAR exploration.