Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-2-amino-2-(thiophen-3-yl)acetic acid | 1194-86-1 | MFCD00079614 | 98.0% | 157.19 | C6H7NO2S | 100 MG
SDP

Catalog No. 5000299905
Encompass_Preferred

(R)-2-Amino-2-(thiophen-3-yl)acetic acid is the R-enantiomer of a thiophene-substituted glycine derivative used as a chiral building block and research reagent. Supplied as a solid with high chemical and enantiomeric purity, it is suitable for incorporation into peptide synthesis, stereospecific transformations, and biochemical studies where a thiophene-containing amino acid is required.

  • High purity (≥98.0%).
  • Molecular weight 157.19 g/mol.
  • Chemical formula C6H7NO2S.
  • Solid, white to off-white appearance.
  • Available in research sizes (100 mg-5 g).
  • Suitable as a chiral building block for synthesis and peptide incorporation.
  • Accompanied by data sheet, certificate of analysis, and safety data sheet.

Catalog No. 50-002-99905 Supplier Medchemexpress LLC Supplier No. HYW017254100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-2-Amino-2-(thiophen-3-yl)acetic acid is the R-enantiomer of a thiophene-substituted glycine derivative used as a chiral building block and research reagent. Supplied as a solid with high chemical and enantiomeric purity, it is suitable for incorporation into peptide synthesis, stereospecific transformations, and biochemical studies where a thiophene-containing amino acid is required.

  • High purity (≥98.0%).
  • Molecular weight 157.19 g/mol.
  • Chemical formula C6H7NO2S.
  • Solid, white to off-white appearance.
  • Available in research sizes (100 mg-5 g).
  • Suitable as a chiral building block for synthesis and peptide incorporation.
  • Accompanied by data sheet, certificate of analysis, and safety data sheet.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.