Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-2-amino-2-(thiophen-3-yl)acetic acid | 1194-86-1 | MFCD00079614 | 98.0% | 157.19 g·mol⁻1 | C6H7NO2S | 250 MG
SDP

Catalog No. 5000303644
Encompass_Preferred

(R)-2-Amino-2-(thiophen-3-yl)acetic acid is the R-enantiomer of a thiophenyl-substituted glycine derivative supplied as a solid for research use. It offers high chemical purity and is intended for synthetic and medicinal chemistry applications requiring enantiomerically defined amino acid building blocks.

  • High purity (98.0%).
  • Molecular weight 157.19 g·mol⁻1.
  • Chemical formula C6H7NO2S.
  • Available in multiple sizes including 250 mg.
  • Suitable for synthetic and medicinal chemistry applications.
  • Enantiomerically defined R-configuration for stereochemical studies.
  • Powder stability under recommended low-temperature storage.

Catalog No. 50-003-03644 Supplier Medchemexpress LLC Supplier No. 250MGHYW017254
May include imposed supplier surcharges.
Add to Cart
Add to Cart

(R)-2-Amino-2-(thiophen-3-yl)acetic acid is the R-enantiomer of a thiophenyl-substituted glycine derivative supplied as a solid for research use. It offers high chemical purity and is intended for synthetic and medicinal chemistry applications requiring enantiomerically defined amino acid building blocks.

  • High purity (98.0%).
  • Molecular weight 157.19 g·mol⁻1.
  • Chemical formula C6H7NO2S.
  • Available in multiple sizes including 250 mg.
  • Suitable for synthetic and medicinal chemistry applications.
  • Enantiomerically defined R-configuration for stereochemical studies.
  • Powder stability under recommended low-temperature storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.