Learn More
Medchemexpress LLC (R)-2-Amino-3-(2-fluorophenyl)propanoic acid | 97731-02-7 | 99.58% | 183.18 | 5 G
SDP
Catalog No. 5000453938
Encompass_Preferred
(R)-2-Amino-3-(2-fluorophenyl)propanoic acid is a phenylalanine derivative. Amino acids and amino acid derivatives are commercially used as ergogenic supplements, influencing the secretion of anabolic hormones, fuel supply during exercise, mental performance during stress-related tasks, and preventing exercise-induced muscle damage. They are recognized to be beneficial as ergogenic dietary substances.
  • Phenylalanine derivative
  • Influences secretion of anabolic hormones
  • Supplies fuel during exercise
  • Enhances mental performance during stress-related tasks
  • Prevents exercise-induced muscle damage
Catalog No. 50-004-53938 Supplier Medchemexpress LLC Supplier No. HYW0100285G
(R)-2-Amino-3-(2-fluorophenyl)propanoic acid is a phenylalanine derivative. Amino acids and amino acid derivatives are commercially used as ergogenic supplements, influencing the secretion of anabolic hormones, fuel supply during exercise, mental performance during stress-related tasks, and preventing exercise-induced muscle damage. They are recognized to be beneficial as ergogenic dietary substances.
  • Phenylalanine derivative
  • Influences secretion of anabolic hormones
  • Supplies fuel during exercise
  • Enhances mental performance during stress-related tasks
  • Prevents exercise-induced muscle damage