Learn More
Medchemexpress LLC (R)-2-amino-3-(furan-2-yl)propanoic acid | 110772-46-8 | MFCD01860879 | ≥97.0% | 155.15 g/mol | C7H9NO3 | 10 G
SDP
Catalog No. 5000210024
Encompass_Preferred
(R)-2-amino-3-(furan-2-yl)propanoic acid is the R-enantiomer of a furan-substituted alanine derivative supplied as a solid for research use. It has CAS number 110772-46-8, molecular formula C7H9NO3, and molecular weight 155.15 g/mol. Typical purity is ≥97.0%, and it is available in multiple package sizes including 10 G.
  • Chiral amino acid derivative useful as a building block in synthesis.
  • High purity (≥97.0%) suitable for biochemical research.
  • Available as an off-white solid powder in multiple sizes including 10 G.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years).
  • COA, data sheet, and SDS available from the manufacturer.
Catalog No. 50-002-10024 Supplier Medchemexpress LLC Supplier No. HYW05156810G
(R)-2-amino-3-(furan-2-yl)propanoic acid is the R-enantiomer of a furan-substituted alanine derivative supplied as a solid for research use. It has CAS number 110772-46-8, molecular formula C7H9NO3, and molecular weight 155.15 g/mol. Typical purity is ≥97.0%, and it is available in multiple package sizes including 10 G.
  • Chiral amino acid derivative useful as a building block in synthesis.
  • High purity (≥97.0%) suitable for biochemical research.
  • Available as an off-white solid powder in multiple sizes including 10 G.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years).
  • COA, data sheet, and SDS available from the manufacturer.