Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-2-amino-3-(furan-2-yl)propanoic acid | 110772-46-8 | MFCD01860879 | ≥97.0% | 155.15 g/mol | C7H9NO3 | 250 MG
SDP

Catalog No. 5000209604
Encompass_Preferred

(R)-2-Amino-3-(furan-2-yl)propanoic acid is the R-enantiomer of a furan-substituted alanine derivative supplied for research use. It is commonly used as a biochemical building block in studies of amino acid metabolism, receptor interactions, and structure-activity relationships.

  • R-enantiomer of a furan-substituted alanine derivative.
  • CAS number 110772-46-8.
  • Molecular formula C7H9NO3; molecular weight 155.15 g/mol.
  • Purity ≥97.0%.
  • Available in small research pack sizes, including 250 mg.

Catalog No. 50-002-09604 Supplier Medchemexpress LLC Supplier No. 250MGHYW051568
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-2-Amino-3-(furan-2-yl)propanoic acid is the R-enantiomer of a furan-substituted alanine derivative supplied for research use. It is commonly used as a biochemical building block in studies of amino acid metabolism, receptor interactions, and structure-activity relationships.

  • R-enantiomer of a furan-substituted alanine derivative.
  • CAS number 110772-46-8.
  • Molecular formula C7H9NO3; molecular weight 155.15 g/mol.
  • Purity ≥97.0%.
  • Available in small research pack sizes, including 250 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.