Learn More
Medchemexpress LLC (R)-2-amino-3-(m-tolyl)propanoic acid | 114926-39-5 | MFCD05227937 | 97.0% | 179.22 g·mol-1 | C10H13NO2 | 25 G
SDP
Catalog No. 5000304157
Encompass_Preferred
(R)-2-Amino-3-(m-tolyl)propanoic acid is the R-enantiomer of a phenylalanine derivative bearing a meta-methyl (m-tolyl) substituent. Supplied as a solid research reagent (CAS 114926-39-5, MW 179.22 g·mol-1), it is used as an enantiomerically defined amino-acid building block in synthetic chemistry and biochemical studies.
  • Enantiomerically defined R-configuration suitable for stereospecific synthesis.
  • Useful as a synthetic building block for peptide and small-molecule synthesis.
  • Suitable for biochemical and pharmacological research applications.
  • Off-white to white solid form for easy handling and storage.
  • Typical purity around 97.0% as provided by the supplier.
Catalog No. 50-003-04157 Supplier Medchemexpress LLC Supplier No. 25GHYW016547
(R)-2-Amino-3-(m-tolyl)propanoic acid is the R-enantiomer of a phenylalanine derivative bearing a meta-methyl (m-tolyl) substituent. Supplied as a solid research reagent (CAS 114926-39-5, MW 179.22 g·mol-1), it is used as an enantiomerically defined amino-acid building block in synthetic chemistry and biochemical studies.
  • Enantiomerically defined R-configuration suitable for stereospecific synthesis.
  • Useful as a synthetic building block for peptide and small-molecule synthesis.
  • Suitable for biochemical and pharmacological research applications.
  • Off-white to white solid form for easy handling and storage.
  • Typical purity around 97.0% as provided by the supplier.