Learn More
Medchemexpress LLC (R)-2-amino-3-(m-tolyl)propanoic acid | 114926-39-5 | MFCD05227937 | 97.0% | 179.22 g/mol | C10H13NO2 | 10 G
SDP
Catalog No. 5000299362
Encompass_Preferred
(R)-2-Amino-3-(m-tolyl)propanoic acid is the R-enantiomer of a phenylalanine derivative used as a chiral amino acid building block in research. It is employed in medicinal chemistry and peptide synthesis to probe steric and electronic effects, and to support structure-activity relationship studies. The compound is a solid with molecular formula C10H13NO2 and molecular weight 179.22 g/mol.
  • R-enantiomer suitable for stereospecific synthesis.
  • Useful as a chiral building block in peptide synthesis.
  • Supports structure-activity relationship and medicinal chemistry research.
  • Solid form for convenient handling and storage.
  • Molecular formula C10H13NO2; molecular weight 179.22 g/mol.
  • Reported purity approximately 97.0% by manufacturer.
Catalog No. 50-002-99362 Supplier Medchemexpress LLC Supplier No. HYW01654710G
(R)-2-Amino-3-(m-tolyl)propanoic acid is the R-enantiomer of a phenylalanine derivative used as a chiral amino acid building block in research. It is employed in medicinal chemistry and peptide synthesis to probe steric and electronic effects, and to support structure-activity relationship studies. The compound is a solid with molecular formula C10H13NO2 and molecular weight 179.22 g/mol.
  • R-enantiomer suitable for stereospecific synthesis.
  • Useful as a chiral building block in peptide synthesis.
  • Supports structure-activity relationship and medicinal chemistry research.
  • Solid form for convenient handling and storage.
  • Molecular formula C10H13NO2; molecular weight 179.22 g/mol.
  • Reported purity approximately 97.0% by manufacturer.