Learn More
Medchemexpress LLC (R)-2-amino-5-(4-(2-(3,5-difluorophenyl)-2-hydroxyacetamido)-2-methylphenyl)-N-isopropylnicotinamide | 3034479-99-4 | 98.9% | 512.51 g/mol | C24H24F2N4O3 · 1/2 C4H4O4 | 10 MG
SDP
Catalog No. 5000288001
Encompass_Preferred
HC-5404-Fu is a selective PERK inhibitor supplied for preclinical research. It modulates endoplasmic reticulum stress signaling and has shown antitumor activity in preclinical models. The compound is provided with high reported purity and is soluble in DMSO for preparation of stock solutions.
  • Selective PERK inhibitor for preclinical studies.
  • Demonstrated antitumor activity and ER stress pathway modulation.
  • Reported purity 98.9% and molecular weight 512.51 g/mol.
  • Soluble in DMSO; suitable for stock solutions.
Catalog No. 50-002-88001 Supplier Medchemexpress LLC Supplier No. HY157231A10MG
HC-5404-Fu is a selective PERK inhibitor supplied for preclinical research. It modulates endoplasmic reticulum stress signaling and has shown antitumor activity in preclinical models. The compound is provided with high reported purity and is soluble in DMSO for preparation of stock solutions.
  • Selective PERK inhibitor for preclinical studies.
  • Demonstrated antitumor activity and ER stress pathway modulation.
  • Reported purity 98.9% and molecular weight 512.51 g/mol.
  • Soluble in DMSO; suitable for stock solutions.